Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01175052_s1
          See other OLFR873 GE Assays ›
          Gene Symbol:
          Olfr873
          Gene Name:
          olfactory receptor 873
          Gene Aliases:
          MOR145-2
          Chromosome Location:
          Chr.9: 20300211 - 20301170 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01175052_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr873
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146561.1 NP_666772.1 1 60 88
          GenBank mRNA BC139140.1 - 1 143 8862162 88

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900000166.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr873

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1993323 NM_146561.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1993323 BC139140.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 942878 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P101325 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1993323 NM_146561.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1993323 BC139140.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26342098 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26342109 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26342105 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26342090 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26342110 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26342097 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26342114 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26342102 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26342112 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26342096 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26342106 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26342089 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26342100 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26342107 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26342104 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26342088 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26342094 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26342092 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26342115 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26342116 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P101325 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126097 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P101325 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07508679 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07824709 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07508680 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07650238 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07650227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26342085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26342118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26342088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26342096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26342084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26342097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26342102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26342121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26342116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26342112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26342104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26342107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26342120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26342094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26342090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26342082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26342100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26342110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26342089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26342117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26342106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26342124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26342092 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26342085 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26342114 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26342113 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26342102 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26342096 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26342094 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26342117 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26342124 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26342106 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07650236 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26342104 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26342098 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26342082 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26342097 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26342090 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26342088 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26342112 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26342092 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26342118 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26342084 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26342086 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26342120 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26342121 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26342110 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26342107 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26342089 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08131476 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26342100 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26342116 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10583447 - -
          Affymetrix Mouse Gene 1.1 ST Array 10583446 - -
          Affymetrix Mouse Clariom S Array PSR0900001030.mm.1 NM_146561.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900001030.mm.1 NM_146561.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900001030.mm.1 TC0900000166.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900001029.mm.1 TC0900000166.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 300692 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3160657 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3160657 - -
          mouse-6 v1.1 (Illumina) GI_49170043-S - -
          MouseRef-8 v2 Expression BeadChip ILMN_3160657 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3160657 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0