Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01175063_m1
          See other B230323A14RIK GE Assays ›
          Gene Symbol:
          B230323A14Rik
          Gene Name:
          RIKEN cDNA B230323A14 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 69761146 - 69830199 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01175063_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: B230323A14Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040765.1 4 - 5 1120 70
          GenBank mRNA AK038443.1 - 4 - 5 1121 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900000921.mm.1
          This assay targets the same location as RTA transcript cluster(s) TC0800001032.rn

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 193372 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 348601 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 519365 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 943198 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 553244 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 707464 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 621992 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 577556 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 134701 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 964944 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 839070 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 679043 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 130062 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 657190 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 420724 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 909508 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 282958 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 803618 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 126604 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 931438 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 779700 AK046229.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 96755 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 886456 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 101571 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 208430 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 777995 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 445380 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 448556 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 117704 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 174836 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 55731 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 86830 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P172725 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P172725 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06832968 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06833021 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06832988 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06832958 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01964317 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100863 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P172725 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06832988 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26612926 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26612773 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26612910 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26612784 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07792257 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26612932 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26612788 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26612913 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26612771 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26612937 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07469123 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26612930 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26612854 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26612851 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26612779 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26612775 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07951993 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26612826 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26612849 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26612786 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08069340 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26612832 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26612825 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26612776 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26612935 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26612847 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26612791 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26612827 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26612780 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26612848 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26612942 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07727915 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26612855 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26612941 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26612916 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26612940 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26612834 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26612856 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26612829 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26612852 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26612927 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26612789 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07667258 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26612929 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26612831 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26612911 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26612828 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26612859 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07792256 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0900007091.mm.1 NR_040765.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900007091.mm.1 NR_040765.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900007091.mm.1 TC0900000921.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900007090.mm.1 NR_040765.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900007090.mm.1 NR_040765.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900007090.mm.1 TC0900000921.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900007094.mm.1 NR_040765.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900007094.mm.1 NR_040765.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900007094.mm.1 TC0900000921.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900007089.mm.1 NR_040765.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900007089.mm.1 NR_040765.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900007089.mm.1 TC0900000921.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900007092.mm.1 NR_040765.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900007092.mm.1 NR_040765.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900007092.mm.1 TC0900000921.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 865060 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 106100519 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0