Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01175227_s1
          See other OLFR877 GE Assays ›
          Gene Symbol:
          Olfr877
          Gene Name:
          olfactory receptor 877
          Gene Aliases:
          MOR161-5
          Chromosome Location:
          Chr.9: 37854820 - 37855755 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01175227_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr877
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146417.1 NP_666529.1 1 471 82

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr877

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2081765 NM_146417.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2081765 NM_146417.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P152065 NM_146417.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26432657 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26432683 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26432668 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26432665 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26432682 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26432686 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26432658 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26432688 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26432654 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26432692 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26432656 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26432653 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26432680 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26432661 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26432673 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26432669 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26432691 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26432666 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26432667 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26432677 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26432660 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26432684 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26432672 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26432674 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26432676 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26432663 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26432675 - -
          Whole Mouse Genome G4122A A_51_P152065 NM_146417.1 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07913646 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07913629 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07913624 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07913648 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07913645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432708 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432641 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432639 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07913645 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432715 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432650 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432673 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432656 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432703 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432658 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432665 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432684 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432689 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07572327 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07913636 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432654 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08017221 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432661 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432666 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432699 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432697 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432675 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432717 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432683 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432653 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432692 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432669 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432691 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432713 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432680 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432649 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432701 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432693 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432711 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432705 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432716 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432686 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432667 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432676 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432663 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432694 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10584359 - -
          Affymetrix Mouse Gene 1.1 ST Array 10584358 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 786787 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 630050 - -
          Illumina mouse-6 v1.0 expression beadchip 630050 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000630050 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1243598 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1243598 - -
          mouse-6 v1.1 (Illumina) scl37108.1_30-S - -
          Illumina Sentrix mouse-6.v1 bead array 630050 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl37108.1_30-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 630050 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1243598 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1243598 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline