Search
Search

Zfp595| Gene Symbol: Zfp595 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177622.3 | NP_808290.1 | 4 - 5 | 1994 | 88 | |
| GenBank mRNA | AK038563.1 | - | 4 - 5 | 1995 | 88 | |
| AK136304.1 | - | 4 - 5 | 1983 | 89 | ||
| AK154058.1 | - | 4 - 5 | 1973 | 88 | ||
| BC141018.1 | - | 4 - 5 | 2067 | 9055630 | 88 | |
Set Membership: |
|