Search
Search

Kmt2b| Gene Symbol: Kmt2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290573.1 | NP_001277502.1 | 34 - 35 | 7691 | 68 | |
| NM_029274.2 | NP_083550.2 | 34 - 35 | 7664 | 68 | ||
| GenBank mRNA | AB093223.1 | - | 29 - 30 | 4791 | 68 | |
| AB182318.1 | - | 34 - 35 | 7655 | 68 | ||
| AK011392.1 | - | 2 - 3 | 144 | 68 | ||
| BC056344.1 | - | 32 - 33 | 5557 | 68 | ||
| BC062210.1 | - | 7 - 8 | 842 | 68 | ||
Set Membership: |
|