| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Gene 1.0 ST Array |
10583462 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10583461 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10583463 |
- |
- |
| Affymetrix HT MG-430 PM Array Plate |
1431312_PM_at |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10583459 |
- |
- |
| Mouse Genome 430 2.0 Array |
1431312_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1431312_PM_at |
- |
- |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array |
66662_at |
- |
- |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array |
66662_PM_at |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR0900001076.mm.1 |
NR_033729.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900001076.mm.1 |
NR_033729.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900001076.mm.1 |
TC0900000174.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900001071.mm.1 |
NR_033729.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900001071.mm.1 |
NR_033729.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900001071.mm.1 |
TC0900000174.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900001072.mm.1 |
NR_033729.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900001072.mm.1 |
NR_033729.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900001072.mm.1 |
TC0900000174.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900001074.mm.1 |
NR_033729.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900001074.mm.1 |
NR_033729.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900001074.mm.1 |
TC0900000174.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900001070.mm.1 |
NR_033729.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900001070.mm.1 |
NR_033729.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900001070.mm.1 |
TC0900000174.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900001064.mm.1 |
NR_033729.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900001064.mm.1 |
NR_033729.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900001064.mm.1 |
TC0900000174.mm.1 |
MTA TC |
| Mouse Expression Set 430 |
1431312_at |
- |
- |
| Mouse Expression Set 430 |
1431312_PM_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1431312_PM_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1431312_at |
- |
- |