Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01175539_m1
          See other GM4791 GE Assays ›
          Gene Symbol:
          Gm4791
          Gene Name:
          predicted gene 4791
          Gene Aliases:
          D930036J05
          Chromosome Location:
          Chr.9: 46998741 - 47003860 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01175539_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm4791
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001243258.1 NP_001230187.1 1 - 2 301 85

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900000571.mm.1
          This assay targets the same location as RTA transcript cluster(s) TC0800000638.rn

          Set Membership:

          Amplicon length between 71 and 85 Recommended Assay Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P133103 AK086549.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P133103 AK086549.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133103 AK086549.1 mouse_transcriptome
          Agilent Six_ChIP_02_of_03_v2 A_68_P26482970 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26482991 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26483035 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07597296 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08165945 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26482972 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26482962 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26482981 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08146731 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26483046 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26482984 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26483042 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26482966 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26482971 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26483050 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26482965 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08117559 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26482994 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08173859 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26482998 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26483037 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26483036 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26482960 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26482975 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07925126 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07592754 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26482989 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26482999 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26483002 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26483044 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26482969 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26483045 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08165944 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26482961 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08117560 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26482958 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07759995 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26482978 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26482993 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26482987 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26482985 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26483047 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07853358 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26482967 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0900004026.mm.1 NM_001243258.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900004026.mm.1 NM_001243258.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900004026.mm.1 TC0900000571.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900004028.mm.1 NM_001243258.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900004028.mm.1 NM_001243258.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900004028.mm.1 TC0900000571.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900004027.mm.1 NM_001243258.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900004027.mm.1 NM_001243258.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900004027.mm.1 TC0900000571.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline