Search
Search

Psmd8| Gene Symbol: Psmd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026545.3 | NP_080821.3 | 1 - 2 | 401 | 294 | |
| GenBank mRNA | AK002921.1 | - | 1 - 2 | 240 | 294 | |
| AK003436.1 | - | 1 - 2 | 242 | 294 | ||
| AK020132.1 | - | 1 - 2 | 260 | 294 | ||
| AK027973.1 | - | 1 - 2 | 205 | 294 | ||
| AK051257.1 | - | 1 - 2 | 253 | 294 | ||
| AK150493.1 | - | 1 - 2 | 244 | 294 | ||
| AK150982.1 | - | 1 - 2 | 208 | 294 | ||
| AK159216.1 | - | 1 - 2 | 210 | 294 | ||
| AK159795.1 | - | 1 - 2 | 226 | 294 | ||
| AK159926.1 | - | 1 - 2 | 236 | 294 | ||
| AK166718.1 | - | 1 - 2 | 235 | 294 | ||
| AK167948.1 | - | 1 - 2 | 210 | 294 | ||
| AK168886.1 | - | 1 - 2 | 209 | 294 | ||
| BC004075.1 | - | 1 - 2 | 219 | 3591796 | 294 | |
| BC005717.1 | - | 1 - 2 | 129 | 2609781 | 294 | |
| CF732286.1 | - | 1 - 2 | 401 | 294 | ||
Set Membership: |
|