Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01175683_m1
          See other MEGF8 GE Assays ›
          Gene Symbol:
          Megf8
          Gene Name:
          multiple EGF-like-domains 8
          Gene Aliases:
          AW049492, Egfl4, b2b1702Clo, b2b288Clo, m687Ddg, mKIAA0817
          Chromosome Location:
          Chr.7: 25316742 - 25365917 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01175683_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Megf8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001160400.1 NP_001153872.1 40 - 41 7450 58
          GenBank mRNA AK030408.1 - 13 - 14 2175 58
          AK122377.1 - 17 - 18 2938 58
          BC036727.1 - 17 - 18 3092 58
          BC060277.1 - 16 - 17 2630 58
          EU723517.1 - 40 - 41 7450 58

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700000413.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          basic leucine zipper transcription factor DNA-binding transcription factor gene-specific transcriptional regulator

          Gene Ontology Categories:

          Function(s) Process(es)

          embryonic heart tube morphogenesis
          regulation of gene expression
          embryonic limb morphogenesis
          BMP signaling pathway
          limb morphogenesis
          cell migration involved in gastrulation
          embryonic skeletal system morphogenesis
          positive regulation of axon extension involved in axon guidance
          epiboly involved in gastrulation with mouth forming second
          epithelial cilium movement involved in determination of left/right asymmetry
          embryonic heart tube left/right pattern formation
          left/right pattern formation
          determination of heart left/right asymmetry
          determination of digestive tract left/right asymmetry
          craniofacial suture morphogenesis
          fasciculation of sensory neuron axon
          nodal signaling pathway involved in determination of lateral mesoderm left/right asymmetry
          molecular_function
          calcium ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P427934 BC060277.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P427934 NM_001160400.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P427934 AK030408.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P427934 AK122377.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P427934 BC031185.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P427934 BC060277.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P427934 NM_001160400.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P427934 AK030408.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P427934 AK122377.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P427934 BC031185.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 462461 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 712148 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 719175 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 777204 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 275770 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 85732 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 838371 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 638542 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 766366 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 40282 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 389991 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 286121 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 131633 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 682038 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 707802 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 245316 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 127409 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 181915 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 174072 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 567525 AK036174.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 567525 AK038702.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 567525 AK053886.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 567525 AK145048.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 567525 AK193489.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 567525 AK194351.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 567525 AK194496.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 567525 AK195084.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 567525 AK195669.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 567525 AK199337.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 567525 AK201482.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 567525 BC007150.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 567525 NM_010017.3 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 567525 X86073.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 567525 Z34532.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 921813 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 133862 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 697930 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 905324 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 12084 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 6211 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 638399 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 82537 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 297123 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 674359 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 440974 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 564508 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 287080 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 137374 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 1335 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 87835 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 122214 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 168457 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P100974 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P170233 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P129695 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P427934 BC060277.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P427934 NM_001160400.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P427934 AK030408.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P427934 AK122377.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P427934 BC031185.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P427934 BC060277.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P427934 NM_001160400.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P427934 AK030408.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P427934 AK122377.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P427934 BC031185.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P427934 BC060277.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P427934 NM_001160400.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P427934 AK030408.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P427934 AK122377.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P427934 BC031185.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P427934 BC060277.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P427934 NM_001160400.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P427934 AK030408.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P427934 AK122377.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P427934 BC031185.1 mouse_transcriptome
          Agilent Six_ChIP_02_of_03_v2 A_68_P05944298 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009858 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009723 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06453783 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009843 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009767 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009820 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009734 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009749 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009838 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009865 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009853 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009763 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009711 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009722 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06147612 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009848 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009833 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009840 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009716 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009806 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009759 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009758 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009736 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06166392 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06649680 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009708 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009817 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009827 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009821 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009824 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009745 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009847 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009760 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009761 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009747 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009815 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009751 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009862 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009864 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06565770 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009812 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06606443 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009766 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06626013 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05924964 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009811 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009728 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05944308 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009783 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009822 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06202586 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009779 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009770 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009830 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009800 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06003003 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009706 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009866 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009729 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06495888 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009762 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05924966 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06032562 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009829 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06242737 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009836 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009733 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06147604 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009851 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06032560 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009825 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009850 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05944296 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06166395 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009704 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009775 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06147601 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009776 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009859 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009863 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009718 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009727 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009769 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06290607 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06202585 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06606439 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009719 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009765 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009788 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06147608 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06003012 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05944291 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009755 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009730 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06626003 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009732 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06147606 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009855 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06469759 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06290606 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009803 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009872 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10551121 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551147 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551150 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551138 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551143 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551136 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551129 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551137 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551128 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551122 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551140 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551154 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551125 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551134 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551130 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551132 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551149 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551153 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551146 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551133 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551144 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551148 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551131 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551145 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551124 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551127 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551151 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551141 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551152 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551135 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551142 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551123 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551139 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551126 - -
          Affymetrix HT MG-430 PM Array Plate 1460586_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10551120 - -
          Mouse Genome 430 2.0 Array 1460586_at - -
          Mouse Genome 430 2.0 Array 1460586_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 269878_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 269878_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700003217.mm.1 TC0700000413.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700003217.mm.1 NM_001160400.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003217.mm.1 NM_001160400.1 Refseq
          Mouse Expression Set 430 1460586_PM_at - -
          Mouse Expression Set 430 1460586_at - -
          Affymetrix HT Mouse Genome 430B Array 1460586_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1460586_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 914308 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_2685985 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1245455 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline