Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01175781_m1
          See other MRLN GE Assays ›
          Gene Symbol:
          Mrln
          Gene Name:
          myoregulin
          Gene Aliases:
          2310015B20Rik, Mln
          Chromosome Location:
          Chr.10: 70204667 - 70219711 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01175781_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mrln
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK009351.1 - 2 - 3 192 65

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P109434 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P134414 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P199352 NR_038041.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P199352 AK009351.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P199352 NR_038041.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P199352 AK009351.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P134414 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P109434 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P111755 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P134414 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P109434 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27228454 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08351114 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08474773 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27228417 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27228491 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08371296 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27228414 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08399375 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27228487 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27228419 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27228428 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27228456 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27228524 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27228412 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27228437 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27228447 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27228427 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08381945 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08898456 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27228433 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27228407 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27228451 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08422515 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27228438 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27228468 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27228472 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27228450 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27228499 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27228439 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08484341 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27228408 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08474770 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27228441 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08465086 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27228476 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27228410 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27228446 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27228495 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27228493 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27228479 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08217537 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08741011 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27228523 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27228404 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27228420 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27228494 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27228496 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27228453 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27228436 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27228434 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27228416 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27228431 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27228452 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27228470 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08371294 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08746880 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27228401 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1430036_PM_at - -
          Mouse Genome 430 2.0 Array 1430036_at - -
          Mouse Genome 430 2.0 Array 1430036_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69563_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69563_PM_at - -
          Mouse Expression Set 430 1430036_PM_at - -
          Mouse Expression Set 430 1430036_at - -
          Affymetrix HT Mouse Genome 430B Array 1430036_at - -
          Affymetrix HT Mouse Genome 430B Array 1430036_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_2737747 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline