Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01176008_m1
          See other TYSND1 GE Assays ›
          Gene Symbol:
          Tysnd1
          Gene Name:
          trypsin domain containing 1
          Gene Aliases:
          1300019N10Rik
          Chromosome Location:
          Chr.10: 61695497 - 61702773 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01176008_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tysnd1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001272090.1 NP_001259019.1 3 - 4 1509 76
          NM_001272091.1 NP_001259020.1 2 - 3 1419 76
          NM_027912.1 NP_082188.1 3 - 4 1550 76
          GenBank mRNA AI181257.1 - 3 - 4 312 76
          AK005069.1 - 3 - 4 1551 76
          AK154513.1 - 3 - 4 1533 76
          BC119320.1 - 3 - 4 1528 8734069 76

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 Protease 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          protease serine protease protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          protein processing
          regulation of fatty acid beta-oxidation
          protein homooligomerization
          protease binding
          serine-type endopeptidase activity
          peptidase activity
          serine-type peptidase activity
          hydrolase activity
          identical protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P517012 AK005069.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P517012 AK154513.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P517012 NM_027912.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P517012 AK005069.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P517012 AK154513.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P517012 NM_027912.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 827484 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 435238 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 373911 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 327545 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 164827 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P131452 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P517012 AK005069.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P517012 AK154513.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P517012 NM_027912.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P517012 AK005069.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P517012 AK154513.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P517012 NM_027912.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02138344 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P131452 - -
          Whole Mouse Genome G4122A A_51_P517012 AK005069.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P517012 AK154513.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P517012 NM_027912.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P517012 AK005069.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P517012 AK154513.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P517012 NM_027912.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P131452 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10363523 - -
          Affymetrix Mouse Gene 1.0 ST Array 10363525 - -
          Affymetrix Mouse Gene 1.0 ST Array 10363526 - -
          Affymetrix Mouse Gene 1.0 ST Array 10363524 - -
          Affymetrix HT MG-430 PM Array Plate 1441856_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1428689_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1428690_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10363522 - -
          Mouse Genome 430 2.0 Array 1428689_PM_at - -
          Mouse Genome 430 2.0 Array 1428690_at - -
          Mouse Genome 430 2.0 Array 1428689_at - -
          Mouse Genome 430 2.0 Array 1441856_PM_x_at - -
          Mouse Genome 430 2.0 Array 1428690_PM_at - -
          Mouse Genome 430 2.0 Array 1441856_x_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71767_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71767_PM_at - -
          Mouse Expression Set 430 1428690_at - -
          Mouse Expression Set 430 1428689_at - -
          Mouse Expression Set 430 1428689_PM_at - -
          Mouse Expression Set 430 1428690_PM_at - -
          Mouse Expression Set 430 1441856_x_at - -
          Mouse Expression Set 430 1441856_PM_x_at - -
          Murine Genome U74v2 Set 104164_at - -
          Murine Genome U74v2 Set 104164_PM_at - -
          Murine Genome U74v2 Set 167623_PM_r_at - -
          Murine Genome U74v2 Set 167718_PM_r_at - -
          Murine Genome U74v2 Set 167623_r_at - -
          Murine Genome U74v2 Set 167718_r_at - -
          Mouse Genome 430A 2.0 Array 1428690_at - -
          Mouse Genome 430A 2.0 Array 1428689_at - -
          Mouse Genome 430A 2.0 Array 1428690_PM_at - -
          Mouse Genome 430A 2.0 Array 1428689_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1428690_at - -
          Affymetrix HT Mouse Genome 430A Array 1428690_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1428689_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1428689_at - -
          Affymetrix HT Mouse Genome 430B Array 1441856_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1441856_PM_x_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 820860 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2940273 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1237212 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1237212 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline