Search
Search

AW551984| Gene Symbol: AW551984 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199556.1 | NP_001186485.1 | 12 - 13 | 1593 | 70 | |
| NM_178737.5 | NP_848852.1 | 12 - 13 | 1583 | 70 | ||
| GenBank mRNA | AK028282.1 | - | 12 - 13 | 1557 | 70 | |
| AK049098.1 | - | 12 - 13 | 1584 | 70 | ||
| AK082321.1 | - | 12 - 13 | 1566 | 70 | ||
| BC061211.1 | - | 11 - 12 | 1279 | 30250574 | 70 | |
Set Membership: |
|