Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01176494_g1
          See other CYP2T4 GE Assays ›
          Gene Symbol:
          Cyp2t4
          Gene Name:
          cytochrome P450, family 2, subfamily t, polypeptide 4
          Gene Aliases:
          Gm1448, Gm1450
          Chromosome Location:
          Chr.7: 27153714 - 27158973 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01176494_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cyp2t4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001100184.1 NP_001093654.1 9 - 10 1332 95

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700000450.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          oxidoreductase oxygenase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          xenobiotic metabolic process
          biological_process
          epoxygenase P450 pathway
          exogenous drug catabolic process
          oxidation-reduction process
          molecular_function
          arachidonic acid epoxygenase activity
          steroid hydroxylase activity
          oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, reduced flavin or flavoprotein as one donor, and incorporation of one atom of oxygen
          oxygen binding
          heme binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2007353 NM_001100184.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 666034 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 89453 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 15695 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2007353 NM_001100184.1 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25015765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25015853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25015794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25015870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25015877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25015842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25015832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25015841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25015798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25015868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25015768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25015770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25015773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25015785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25015833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25015758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25015796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25015792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25015783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25015752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25015748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25015836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25015834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25015871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25015844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25015826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25015830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25015878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25015839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25015863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25015763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25015873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25015828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25015843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25015766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25015847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25015800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25015850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25015835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25015777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25015879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25015856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25015822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25015781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25015854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25015789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25015859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25015849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25015852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25015750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25015876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25015787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25015861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25015776 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25015794 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25015798 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25015787 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25015779 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25015759 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06211383 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25015758 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25015761 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25015768 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25015764 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06219189 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06219185 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25015795 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25015762 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25015780 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25015773 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25015763 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25015777 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25015770 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06187683 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25015783 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25015800 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25015765 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25015785 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25015803 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25015776 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06189929 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25015781 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25015767 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10551310 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551312 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551305 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551306 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551309 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551313 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551308 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551307 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551311 - -
          Affymetrix Mouse Gene 1.1 ST Array 10551304 - -
          Affymetrix Mouse Clariom S Array PSR0700003540.mm.1 TC0700000450.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700003540.mm.1 NM_001100184.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003540.mm.1 NM_001100184.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 508988 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1254560 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline