Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01176552_g1
          See other HAUS5 GE Assays ›
          Gene Symbol:
          Haus5
          Gene Name:
          HAUS augmin-like complex, subunit 5
          Gene Aliases:
          2310022K01Rik
          Chromosome Location:
          Chr.7: 30653708 - 30664994 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01176552_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Haus5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027999.1 NP_082275.1 14 - 15 1333 100
          GenBank mRNA AK009476.1 - 14 - 15 1313 100
          AK129228.1 - 10 - 11 2601 100
          AK153920.1 - 14 - 15 1315 100
          BC023723.1 - 14 - 15 1283 100
          BC089002.1 - 14 - 15 1302 100
          BC118934.1 - 14 - 15 1308 40090953 100
          BC118935.1 - 14 - 15 1308 40090954 100

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700002773.mm.1

          Set Membership:

          Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          cell cycle
          mitotic nuclear division
          spindle assembly
          centrosome organization
          cell division
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1973178 AK129228.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1973178 BC048724.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1973178 BC089002.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1973178 AK153920.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1973178 AK194141.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 189905 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 631437 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 171417 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 512831 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 556868 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 507313 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 147867 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P127517 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P172633 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1973178 AK129228.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1973178 BC048724.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1973178 BC089002.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1973178 AK153920.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1973178 AK194141.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P316545 BC118935.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P316545 AK129228.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P316545 BC048724.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P316545 BC089002.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P316545 BC118934.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P124008 AK129228.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P124008 BC048724.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P124008 BC089002.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P124008 AK153920.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P124008 AK194141.1 mouse_transcriptome
          Agilent Six_ChIP_02_of_03_v2 A_68_P06491807 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25032791 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25032813 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06173132 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25032793 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25032794 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25032825 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25032795 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06173137 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25032828 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25032809 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06045176 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25032831 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25032822 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25032806 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25032804 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25032799 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06045177 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06491805 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06491809 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25032835 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25032810 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25032837 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06172848 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25032832 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25032815 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25032807 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06360383 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10562086 - -
          Affymetrix Mouse Gene 1.0 ST Array 10562090 - -
          Affymetrix Mouse Gene 1.0 ST Array 10562089 - -
          Affymetrix Mouse Gene 1.0 ST Array 10352357 - -
          Affymetrix Mouse Gene 1.0 ST Array 10562092 - -
          Affymetrix Mouse Gene 1.0 ST Array 10562088 - -
          Affymetrix Mouse Gene 1.0 ST Array 10562091 - -
          Affymetrix Mouse Gene 1.0 ST Array 10562085 - -
          Affymetrix Mouse Gene 1.0 ST Array 10562087 - -
          Affymetrix HT MG-430 PM Array Plate 1452715_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10352356 - -
          Affymetrix Mouse Gene 1.1 ST Array 10562084 - -
          Mouse Genome 430 2.0 Array 1452715_PM_at - -
          Mouse Genome 430 2.0 Array 1452715_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71909_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71909_at - -
          Affymetrix Mouse Clariom S Array PSR0700024731.mm.1 TC0700002773.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024731.mm.1 NM_027999.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024731.mm.1 NM_027999.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024732.mm.1 TC0700002773.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024732.mm.1 NM_027999.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024732.mm.1 NM_027999.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024741.mm.1 TC0700002773.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024741.mm.1 NM_027999.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024741.mm.1 NM_027999.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024735.mm.1 TC0700002773.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024735.mm.1 NM_027999.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024735.mm.1 NM_027999.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024738.mm.1 TC0700002773.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024738.mm.1 NM_027999.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024738.mm.1 NM_027999.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024745.mm.1 TC0700002773.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024745.mm.1 NM_027999.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024745.mm.1 NM_027999.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024751.mm.1 TC0700002773.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024751.mm.1 NM_027999.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024751.mm.1 NM_027999.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024759.mm.1 TC0700002773.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024759.mm.1 NM_027999.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024759.mm.1 NM_027999.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024739.mm.1 TC0700002773.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024739.mm.1 NM_027999.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024739.mm.1 NM_027999.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024749.mm.1 TC0700002773.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024749.mm.1 NM_027999.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024749.mm.1 NM_027999.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024757.mm.1 TC0700002773.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024757.mm.1 NM_027999.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024757.mm.1 NM_027999.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024760.mm.1 TC0700002773.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024760.mm.1 NM_027999.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024760.mm.1 NM_027999.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024762.mm.1 TC0700002773.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024762.mm.1 NM_027999.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024762.mm.1 NM_027999.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024754.mm.1 TC0700002773.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024754.mm.1 NM_027999.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024754.mm.1 NM_027999.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024761.mm.1 TC0700002773.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024761.mm.1 NM_027999.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024761.mm.1 NM_027999.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024766.mm.1 TC0700002773.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024766.mm.1 NM_027999.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024766.mm.1 NM_027999.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024733.mm.1 TC0700002773.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024733.mm.1 NM_027999.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024733.mm.1 NM_027999.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024736.mm.1 TC0700002773.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024736.mm.1 NM_027999.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024736.mm.1 NM_027999.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024758.mm.1 TC0700002773.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024758.mm.1 NM_027999.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024758.mm.1 NM_027999.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024747.mm.1 TC0700002773.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024747.mm.1 NM_027999.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024747.mm.1 NM_027999.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024743.mm.1 TC0700002773.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024743.mm.1 NM_027999.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024743.mm.1 NM_027999.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024753.mm.1 TC0700002773.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024753.mm.1 NM_027999.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024753.mm.1 NM_027999.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024755.mm.1 TC0700002773.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024755.mm.1 NM_027999.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024755.mm.1 NM_027999.1 Refseq
          Mouse Expression Set 430 1452715_at - -
          Mouse Expression Set 430 1452715_PM_at - -
          Murine Genome U74v2 Set 97371_PM_at - -
          Murine Genome U74v2 Set 97371_at - -
          Mouse Genome 430A 2.0 Array 1452715_PM_at - -
          Mouse Genome 430A 2.0 Array 1452715_at - -
          Affymetrix HT Mouse Genome 430A Array 1452715_at - -
          Affymetrix HT Mouse Genome 430A Array 1452715_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 400260 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 100360280 - -
          Illumina mouse-6 v1.0 expression beadchip 100360280 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1242680 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1235574 - -
          Illumina Sentrix mouse-6.v1 bead array 100360280 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 100360280 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1242680 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline