Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700004758.mm.1 |
TC0700000557.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700004758.mm.1 |
NM_001081114.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700004758.mm.1 |
NM_001081114.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700004753.mm.1 |
TC0700000557.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700004753.mm.1 |
NM_001081114.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700004753.mm.1 |
NM_001081114.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700004760.mm.1 |
TC0700000557.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700004760.mm.1 |
NM_001081114.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700004760.mm.1 |
NM_001081114.1 |
Refseq |