Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01176664_g1
          See other POLR2I GE Assays ›
          Gene Symbol:
          Polr2i
          Gene Name:
          polymerase (RNA) II (DNA directed) polypeptide I
          Gene Aliases:
          2810002B19Rik
          Chromosome Location:
          Chr.7: 30232074 - 30233387 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01176664_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Polr2i
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027259.1 NP_081535.1 5 - 6 590 123
          GenBank mRNA BC062812.1 - 5 - 6 590 3466417 123

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700000554.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          DNA-directed RNA polymerase RNA metabolism protein nucleic acid metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          maintenance of transcriptional fidelity during DNA-templated transcription elongation from RNA polymerase II promoter
          transcription-coupled nucleotide-excision repair
          transcription, DNA-templated
          transcription from RNA polymerase II promoter
          transcription initiation from RNA polymerase II promoter
          nucleic acid binding
          DNA binding
          DNA-directed RNA polymerase activity
          zinc ion binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25030467 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25030471 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25030463 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25030466 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25030461 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25030469 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2135601 BC062812.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2135601 NM_027259.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1967720 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 215043 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 519237 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1967720 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2135601 BC062812.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2135601 NM_027259.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P246215 AK012635.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P246215 AK012635.1 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06221487 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05932566 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05932570 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25030461 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25030467 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25030469 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25030471 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25030466 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25030463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030493 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030507 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030528 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030524 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06605819 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06476906 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030474 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06476920 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030516 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030515 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030498 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06537702 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06354149 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030475 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030525 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030463 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030492 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030488 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06048947 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06605824 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05932579 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030479 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030461 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030471 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030469 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06476914 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05932580 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030531 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030518 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030500 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06431245 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030491 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06605827 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030489 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030521 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030470 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030476 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030482 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030523 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030520 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030529 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030466 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030501 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030508 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030465 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030494 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06605825 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10551843 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551845 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551846 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551842 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551847 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551844 - -
          Affymetrix HT MG-430 PM Array Plate 1428494_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10551841 - -
          Mouse Genome 430 2.0 Array 1428494_a_at - -
          Mouse Genome 430 2.0 Array 1428494_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69920_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69920_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700004736.mm.1 TC0700000554.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700004736.mm.1 NM_027259.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700004736.mm.1 NM_027259.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700004739.mm.1 TC0700000554.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700004739.mm.1 NM_027259.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700004739.mm.1 NM_027259.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700004738.mm.1 TC0700000554.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700004738.mm.1 NM_027259.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700004738.mm.1 NM_027259.1 Refseq
          Mouse Expression Set 430 1428494_PM_a_at - -
          Mouse Expression Set 430 1428494_a_at - -
          Murine Genome U74v2 Set 96306_at - -
          Murine Genome U74v2 Set 96306_PM_at - -
          Mouse Genome 430A 2.0 Array 1428494_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1428494_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 782574 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4060400 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2666438 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2666438 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2666438 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline