Search
Search

Fam219b| Gene Symbol: Fam219b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166364.1 | NP_001159836.1 | 2 - 3 | 394 | 61 | |
| NM_175273.4 | NP_780482.1 | 2 - 3 | 394 | 61 | ||
| GenBank mRNA | AK028365.1 | - | 2 - 3 | 395 | 61 | |
| AK185896.1 | - | 1 - 2 | 55 | 61 | ||
| BC111883.1 | - | 2 - 3 | 359 | 40060305 | 61 | |
| BC112403.1 | - | 2 - 3 | 359 | 40060304 | 61 | |
Set Membership: |
|