Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01177345_m1
          See other CCDC105 GE Assays ›
          Gene Symbol:
          Ccdc105
          Gene Name:
          coiled-coil domain containing 105
          Gene Aliases:
          4931413A09Rik
          Chromosome Location:
          Chr.10: 78746924 - 78753067 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01177345_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ccdc105
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027630.1 NP_081906.1 6 - 7 1346 98

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000002461.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Recommended Assay Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2111108 NM_027630.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 848753 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 372539 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 117888 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2111108 NM_027630.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P462569 AK016452.1 mouse_transcriptome
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275493 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08339947 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275473 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08218154 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08856971 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275483 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275488 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08814794 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275492 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08971057 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275470 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08390982 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08857495 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08390986 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08857488 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08390983 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08715047 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275489 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275477 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275469 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275475 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08569879 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275467 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275494 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275471 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275505 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275495 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275502 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08625203 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275486 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08505775 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1453702_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10370530 - -
          Mouse Genome 430 2.0 Array 1453702_at - -
          Mouse Genome 430 2.0 Array 1453702_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70976_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70976_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1000017936.mm.1 NM_027630.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000017936.mm.1 NM_027630.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000017936.mm.1 TC1000002461.mm.1 MTA TC
          Mouse Expression Set 430 1453702_at - -
          Mouse Expression Set 430 1453702_PM_at - -
          Murine Genome U74v2 Set 168247_PM_f_at - -
          Murine Genome U74v2 Set 168247_f_at - -
          Murine Genome U74v2 Set 168246_at - -
          Murine Genome U74v2 Set 168246_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1453702_at - -
          Affymetrix HT Mouse Genome 430B Array 1453702_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 499676 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2667857 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2667857 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0