| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0900002091.mm.1 |
NM_001101597.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900002091.mm.1 |
NM_001101597.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900002091.mm.1 |
TC0900000278.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900002090.mm.1 |
NM_001101597.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900002090.mm.1 |
NM_001101597.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900002090.mm.1 |
TC0900000278.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900002093.mm.1 |
NM_001101597.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900002093.mm.1 |
NM_001101597.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900002093.mm.1 |
TC0900000278.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900002092.mm.1 |
NM_001101597.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900002092.mm.1 |
NM_001101597.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900002092.mm.1 |
TC0900000278.mm.1 |
MTA TC |