Search
Search

Olfr957| Gene Symbol: Olfr957 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146745.2 | NP_666956.2 | 1 | 466 | 81 | |
| GenBank mRNA | BC106794.2 | - | 1 | 462 | 81 | |
| BC114587.1 | - | 1 | 465 | 81 | ||
| BC115839.1 | - | 1 | 462 | 81 | ||
Set Membership: |
|