| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100012801.mm.1 |
NM_001078167.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012801.mm.1 |
NM_173374.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012801.mm.1 |
TC1100001359.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100012807.mm.1 |
NM_001078167.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012807.mm.1 |
NM_173374.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012807.mm.1 |
TC1100001359.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100012796.mm.1 |
NM_001078167.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012796.mm.1 |
NM_173374.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012796.mm.1 |
TC1100001359.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100012803.mm.1 |
NM_001078167.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012803.mm.1 |
NM_173374.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012803.mm.1 |
TC1100001359.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100012795.mm.1 |
NM_001078167.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012795.mm.1 |
NM_173374.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012795.mm.1 |
TC1100001359.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100012798.mm.1 |
NM_001078167.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012798.mm.1 |
NM_173374.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012798.mm.1 |
TC1100001359.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100012808.mm.1 |
NM_001078167.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012808.mm.1 |
NM_173374.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012808.mm.1 |
TC1100001359.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100012799.mm.1 |
NM_001078167.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012799.mm.1 |
NM_173374.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012799.mm.1 |
TC1100001359.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100012809.mm.1 |
NM_001078167.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012809.mm.1 |
NM_173374.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012809.mm.1 |
TC1100001359.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100012797.mm.1 |
NM_001078167.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012797.mm.1 |
NM_173374.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012797.mm.1 |
TC1100001359.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100012802.mm.1 |
NM_001078167.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012802.mm.1 |
NM_173374.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012802.mm.1 |
TC1100001359.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100012804.mm.1 |
NM_001078167.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012804.mm.1 |
NM_173374.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012804.mm.1 |
TC1100001359.mm.1 |
MTA TC |