Search
Search

Kirrel3| Gene Symbol: Kirrel3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190911.1 | NP_001177840.1 | 7 - 8 | 1225 | 65 | |
| NM_001190912.1 | NP_001177841.1 | 6 - 7 | 1147 | 65 | ||
| NM_001190913.1 | NP_001177842.1 | 7 - 8 | 1225 | 65 | ||
| NM_001190914.1 | NP_001177843.1 | 8 - 9 | 1326 | 65 | ||
| NM_026324.3 | NP_080600.1 | 7 - 8 | 1225 | 65 | ||
| GenBank mRNA | AK034142.1 | - | 7 - 8 | 1162 | 65 | |
| AK038385.1 | - | 7 - 8 | 976 | 65 | ||
| AK045373.1 | - | 7 - 8 | 1130 | 65 | ||
| AK079332.1 | - | 8 - 9 | 1264 | 65 | ||
| AK171866.1 | - | 4 - 5 | 576 | 65 | ||
| AY169782.1 | - | 7 - 8 | 1225 | 65 | ||
| BC063072.1 | - | 6 - 7 | 942 | 5695003 | 65 | |
Set Membership: |
|