| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0900021135.mm.1 |
NM_001025612.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900021135.mm.1 |
NM_001025612.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900021135.mm.1 |
TC0900002521.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900021134.mm.1 |
NM_001025612.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900021134.mm.1 |
NM_001025612.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900021134.mm.1 |
TC0900002521.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900021133.mm.1 |
NM_001025612.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900021133.mm.1 |
NM_001025612.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900021133.mm.1 |
TC0900002521.mm.1 |
MTA TC |