Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01178961_m1
          See other ZFP712 GE Assays ›
          Gene Symbol:
          Zfp712
          Gene Name:
          zinc finger protein 712
          Gene Aliases:
          4921504N20Rik, mszf31, mszf89
          Chromosome Location:
          Chr.13: 67038594 - 67061170 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01178961_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp712
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001166218.1 NP_001159690.1 3 - 4 336 75
          GenBank mRNA AK019536.1 - 3 - 4 317 75
          BB642289.1 - 3 - 4 336 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300002239.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Recommended Assay Mus musculus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription, DNA-templated
          transcription factor activity, sequence-specific DNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2147126 NM_001166218.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 817076 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 115323 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 218071 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 594806 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 887849 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 530265 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 908022 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 961264 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 542405 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 354073 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 965636 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 161692 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 170648 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 150853 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2147126 NM_001166218.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P260020 AK019536.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P260020 NM_001166218.1 mouse_transcriptome
          Agilent Six_ChIP_03_of_03_v2 A_68_P10449581 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29107192 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10636603 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29107189 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29107196 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29107199 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29107193 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29107202 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29107200 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29107198 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29107203 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29107185 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29107205 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1430212_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1440778_PM_x_at - -
          Mouse Genome 430 2.0 Array 1430212_PM_at - -
          Mouse Genome 430 2.0 Array 1440778_PM_x_at - -
          Mouse Genome 430 2.0 Array 1440778_x_at - -
          Mouse Genome 430 2.0 Array 1430212_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 78251_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 78251_at - -
          Affymetrix Mouse Clariom S Array PSR1300014517.mm.1 NM_001166218.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300014517.mm.1 NM_001166218.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300014517.mm.1 TC1300002239.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300014518.mm.1 NM_001166218.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300014518.mm.1 NM_001166218.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300014518.mm.1 TC1300002239.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300014519.mm.1 NM_001166218.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300014519.mm.1 NM_001166218.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300014519.mm.1 TC1300002239.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300014516.mm.1 NM_001166218.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300014516.mm.1 NM_001166218.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300014516.mm.1 TC1300002239.mm.1 MTA TC
          Mouse Expression Set 430 1430212_PM_at - -
          Mouse Expression Set 430 1430212_at - -
          Mouse Expression Set 430 1440778_x_at - -
          Mouse Expression Set 430 1440778_PM_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1440778_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1440778_PM_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1430212_at - -
          Affymetrix HT Mouse Genome 430B Array 1430212_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 305255 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2644445 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2644445 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline