Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01179110_m1
          See other FBXO27 GE Assays ›
          Gene Symbol:
          Fbxo27
          Gene Name:
          F-box protein 27
          Gene Aliases:
          E130008B10Rik, FBG5, Gm161
          Chromosome Location:
          Chr.7: 28692779 - 28699338 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01179110_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fbxo27
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001163702.1 NP_001157174.1 3 - 4 712 81
          NM_207238.3 NP_997121.1 4 - 5 707 81
          GenBank mRNA AK053292.1 - 4 - 5 709 81
          AK140275.1 - 3 - 4 710 81
          AK158463.1 - 3 - 4 713 81
          AK169614.1 - 3 - 4 797 81
          BC075656.1 - 3 - 4 675 30540609 81

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700000507.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fbxo27

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          glycoprotein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25023124 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25023125 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25023126 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25023123 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2170847 BC075656.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2170847 BQ033399.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2170847 AK140275.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2170847 AK158463.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2170847 AK169614.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 785051 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 287603 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 657066 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 87206 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P156575 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2170847 BC075656.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2170847 BQ033399.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2170847 AK140275.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2170847 AK158463.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2170847 AK169614.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023136 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023137 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023135 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023120 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023133 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023109 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023128 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023125 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023127 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023129 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023126 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023111 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023134 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023132 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023123 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023117 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023115 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023131 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023124 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023114 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06150141 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P156575 - -
          Whole Mouse Genome G4122A A_51_P265173 BC075656.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P265173 BQ033399.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P265173 AK140275.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P265173 AK158463.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P265173 AK169614.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P103329 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P156575 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06646652 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06646650 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06646644 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25023125 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25023126 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25023124 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25023123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023115 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023117 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023145 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023116 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023184 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023111 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023134 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023156 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023123 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05958593 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023117 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023180 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023138 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023129 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023143 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06263737 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023154 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023185 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06646640 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023127 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023183 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06397014 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023125 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023155 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023152 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06407350 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023126 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023132 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023115 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06646649 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06419066 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06188535 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023182 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06212686 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06139648 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023112 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05990401 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023148 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023146 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06469689 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10551539 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551538 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551536 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551535 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551540 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551537 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551541 - -
          Affymetrix HT MG-430 PM Array Plate 1447296_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1434122_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10551534 - -
          Mouse Genome 430 2.0 Array 1447296_at - -
          Mouse Genome 430 2.0 Array 1447296_PM_at - -
          Mouse Genome 430 2.0 Array 1434122_at - -
          Mouse Genome 430 2.0 Array 1434122_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 233040_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 233040_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700004122.mm.1 TC0700000507.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700004122.mm.1 NM_001163702.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700004122.mm.1 NM_001163702.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700004122.mm.1 NM_207238.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700004122.mm.1 NM_207238.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700004124.mm.1 TC0700000507.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700004124.mm.1 NM_001163702.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700004124.mm.1 NM_001163702.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700004124.mm.1 NM_207238.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700004124.mm.1 NM_207238.3 Refseq
          Mouse Expression Set 430 1434122_at - -
          Mouse Expression Set 430 1447296_PM_at - -
          Mouse Expression Set 430 1434122_PM_at - -
          Mouse Expression Set 430 1447296_at - -
          Affymetrix HT Mouse Genome 430B Array 1447296_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1434122_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1434122_at - -
          Affymetrix HT Mouse Genome 430B Array 1447296_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 889639 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4760100 - -
          Illumina mouse-6 v1.0 expression beadchip 4760100 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004760100 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2737738 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2737738 - -
          mouse-6 v1.1 (Illumina) scl0233040.6_239-S - -
          Illumina Sentrix mouse-6.v1 bead array 4760100 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0233040.6_239-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4760100 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2737738 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2737738 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0233040.6_239-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0