Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01179123_m1
          See other SCGB1B2 GE Assays ›
          Gene Symbol:
          Scgb1b2
          Gene Name:
          secretoglobin, family 1B, member 2
          Gene Aliases:
          Abpa2, Abph, Apbh, LGP, Mja1l, Scgb1b1
          Chromosome Location:
          Chr.7: 31290519 - 31291816 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01179123_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Scgb1b2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_020563.3 NP_065588.1 2 - 3 261 102
          GenBank mRNA AF008595.1 - 2 - 3 262 102
          BC115490.1 - 2 - 3 262 40100845 102
          BC115491.1 - 2 - 3 262 40100846 102

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700002793.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Scgb1b2

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2152222 NM_020563.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2152222 AF008595.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2152222 BC115490.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2152222 BC115491.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2152222 BX629566.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P114930 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2152222 NM_020563.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2152222 AF008595.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2152222 BC115490.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2152222 BC115491.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2152222 BX629566.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25035745 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25035753 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25035744 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25035772 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25035751 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25035768 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25035735 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25035771 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25035761 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25035740 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25035749 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25035736 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25035728 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25035775 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25035755 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25035774 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25035759 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25035757 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25035783 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25035738 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25035773 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25035780 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25035754 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25035781 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P114930 - -
          Whole Mouse Genome G4122A A_51_P151211 NM_020563.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P151211 AF008595.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P151211 BC115490.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P151211 BC115491.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P151211 BX629566.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P128338 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P114930 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06552610 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06552609 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06552606 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06552608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25035773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25035740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25035772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25035745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25035735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25035751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25035736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25035781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25035775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25035714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25035755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25035761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25035780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25035757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25035734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25035715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25035738 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06387435 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25035715 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25035753 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25035749 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25035738 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06206537 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25035736 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25035755 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25035722 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25035724 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06552619 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25035714 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25035751 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06387438 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06552613 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25035735 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25035734 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10562282 - -
          Affymetrix Mouse Gene 1.0 ST Array 10562284 - -
          Affymetrix Mouse Gene 1.0 ST Array 10562281 - -
          Affymetrix Mouse Gene 1.0 ST Array 10562283 - -
          Affymetrix HT MG-430 PM Array Plate 1421803_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10562280 - -
          Mouse Genome 430 2.0 Array 1421803_PM_at - -
          Mouse Genome 430 2.0 Array 1421803_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 57426_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 57426_at - -
          Affymetrix Mouse Clariom S Array PSR0700025083.mm.1 TC0700002793.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700025083.mm.1 NM_020563.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700025083.mm.1 NM_020563.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700025084.mm.1 TC0700002793.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700025084.mm.1 NM_020563.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700025084.mm.1 NM_020563.3 Refseq
          Mouse Expression Set 430 1421803_PM_at - -
          Mouse Expression Set 430 1421803_at - -
          Murine Genome U74v2 Set 101621_PM_at - -
          Murine Genome U74v2 Set 101621_at - -
          Mouse Genome 430A 2.0 Array 1421803_PM_at - -
          Mouse Genome 430A 2.0 Array 1421803_at - -
          Affymetrix HT Mouse Genome 430A Array 1421803_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1421803_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 594963 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3850500 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003850500 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1244436 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1244436 - -
          mouse-6 v1.1 (Illumina) scl31494.3.1_32-S - -
          Illumina Sentrix mouse-6.v1 bead array 3850500 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl31494.3.1_32-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3850500 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1244436 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1244436 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline