Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01179173_m1
          See other TMEM136 GE Assays ›
          Gene Symbol:
          Tmem136
          Gene Name:
          transmembrane protein 136
          Gene Aliases:
          AA396355, Gm184
          Chromosome Location:
          Chr.9: 43108650 - 43116714 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01179173_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmem136
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001034863.3 NP_001030035.1 2 - 3 297 85
          GenBank mRNA AK128910.1 - 2 - 3 346 85
          AK158698.1 - 2 - 3 298 85
          BC139046.1 - 2 - 3 284 8862068 85
          BC139047.1 - 2 - 3 284 8862069 85

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900002126.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26459754 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26459756 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26459755 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26459752 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P118772 AK158698.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P118772 NM_001034863.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916051 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 50317 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 519512 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 494969 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 389982 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 158187 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P113243 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P118772 AK158698.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P118772 NM_001034863.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26459755 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26459764 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26459765 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26459782 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26459711 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26459747 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26459715 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26459758 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26459710 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26459720 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26459708 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26459748 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26459752 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26459760 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26459749 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26459722 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26459771 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26459773 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26459712 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26459768 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26459757 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26459766 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26459754 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26459751 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26459745 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26459777 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26459781 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26459743 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26459769 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26459759 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26459742 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26459762 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26459779 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26459717 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P113243 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P113243 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26459755 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26459756 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26459752 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26459754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26459687 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26459742 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26459794 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07743914 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26459712 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26459710 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26459739 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08055931 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26459773 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26459762 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26459769 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08172972 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26459792 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08055922 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07636568 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26459738 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26459777 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26459774 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08112387 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26459787 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26459766 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26459782 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26459735 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07494400 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07914424 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26459733 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26459708 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26459729 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26459741 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26459771 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26459723 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08055935 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26459760 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26459779 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26459770 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26459717 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26459783 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26459726 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26459761 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26459725 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26459711 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26459732 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26459746 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08055918 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07743919 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26459715 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26459724 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26459768 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26459737 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26459759 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26459789 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26459718 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26459781 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26459730 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26459731 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26459751 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10592704 - -
          Affymetrix Mouse Gene 1.0 ST Array 10592703 - -
          Affymetrix Mouse Gene 1.0 ST Array 10592702 - -
          Affymetrix HT MG-430 PM Array Plate 1457701_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10592701 - -
          Mouse Genome 430 2.0 Array 1457701_PM_at - -
          Mouse Genome 430 2.0 Array 1457701_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 235300_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 235300_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0900017684.mm.1 NM_001034863.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900017684.mm.1 NM_001034863.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900017684.mm.1 TC0900002126.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900017683.mm.1 NM_001034863.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900017683.mm.1 NM_001034863.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900017683.mm.1 TC0900002126.mm.1 MTA TC
          Mouse Expression Set 430 1457701_PM_at - -
          Mouse Expression Set 430 1457701_at - -
          Affymetrix HT Mouse Genome 430B Array 1457701_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1457701_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 555247 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_2533407 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2809571 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2809571 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline