Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01179194_m1
          See other CD3E GE Assays ›
          Gene Symbol:
          Cd3e
          Gene Name:
          CD3 antigen, epsilon polypeptide
          Gene Aliases:
          AI504783, CD3, CD3epsilon, T3e
          Chromosome Location:
          Chr.9: 44998735 - 45009662 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01179194_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cd3e
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_007648.4 NP_031674.1 7 - 8 592 121
          GenBank mRNA BC145926.1 - 7 - 8 592 40131229 121
          BC145954.1 - 7 - 8 592 40131257 121
          J02990.1 - 7 - 8 593 121

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900002189.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cd3e

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of T cell anergy
          cell surface receptor signaling pathway
          response to nutrient
          T cell costimulation
          positive regulation of interferon-gamma production
          positive regulation of interleukin-4 production
          positive regulation of T cell proliferation
          negative thymic T cell selection
          positive regulation of interleukin-2 biosynthetic process
          negative regulation of smoothened signaling pathway
          positive regulation of alpha-beta T cell proliferation
          lymphocyte activation
          positive regulation of peptidyl-tyrosine phosphorylation
          positive regulation of calcium-mediated signaling
          T cell receptor signaling pathway
          positive regulation of T cell activation
          apoptotic signaling pathway
          signal transducer activity
          transmembrane signaling receptor activity
          protein binding
          SH3 domain binding
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2156229 J02990.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2156229 NM_007648.4 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 54396 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 749728 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 421618 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 701465 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 774594 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 611228 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 57089 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P108588 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P103901 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2156229 J02990.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2156229 NM_007648.4 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P455681 J02990.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P455681 NM_007648.4 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26470859 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26470865 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26470844 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26470868 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26470846 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26470853 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26470861 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26470843 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26470879 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26470874 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26470864 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26470849 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26470871 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26470841 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26470854 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26470875 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26470847 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26470842 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26470851 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26470852 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26470878 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26470857 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26470855 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26470870 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P108588 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06771027 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P103901 - -
          Whole Mouse Genome G4122A A_51_P455681 J02990.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P455681 NM_007648.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P86404 BC028636.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P86404 BC098236.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P86404 AK041607.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P122005 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P108588 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P103901 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07572180 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07572172 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07572173 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07572171 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07572181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470788 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470872 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26470866 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26470818 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26470811 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26470877 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26470790 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07469361 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26470780 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26470842 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26470812 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07518896 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26470868 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07469351 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26470821 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26470810 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26470851 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07630275 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26470853 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26470874 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26470779 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26470838 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26470861 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26470854 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26470777 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26470882 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07630274 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26470835 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26470834 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07891113 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26470879 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26470823 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07579265 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26470830 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26470849 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26470855 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26470814 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26470857 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26470837 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26470859 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26470785 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26470883 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26470884 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26470840 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26470826 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26470816 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26470880 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26470819 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07681467 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26470784 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26470820 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26470844 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26470865 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07469346 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26470825 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08155388 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07788867 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26470839 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26470864 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26470875 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26470852 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26470787 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26470788 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10593025 - -
          Affymetrix Mouse Gene 1.0 ST Array 10593031 - -
          Affymetrix Mouse Gene 1.0 ST Array 10593029 - -
          Affymetrix Mouse Gene 1.0 ST Array 10593028 - -
          Affymetrix Mouse Gene 1.0 ST Array 10593026 - -
          Affymetrix Mouse Gene 1.0 ST Array 10593030 - -
          Affymetrix Mouse Gene 1.0 ST Array 10593027 - -
          Affymetrix HT MG-430 PM Array Plate 1422105_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10593024 - -
          Mouse Genome 430 2.0 Array 1445748_at - -
          Mouse Genome 430 2.0 Array 1422105_at - -
          Mouse Genome 430 2.0 Array 1445748_PM_at - -
          Mouse Genome 430 2.0 Array 1422105_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12501_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12501_PM_at - -
          Affymetrix Murine Genome 6500A Array Msa12080_at - -
          Affymetrix Mouse Clariom S Array PSR0900018268.mm.1 NM_007648.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900018268.mm.1 NM_007648.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0900018268.mm.1 TC0900002189.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900018267.mm.1 NM_007648.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900018267.mm.1 NM_007648.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0900018267.mm.1 TC0900002189.mm.1 MTA TC
          Mouse Expression Set 430 1422105_PM_at - -
          Mouse Expression Set 430 1422105_at - -
          Murine Genome U74v2 Set 102971_PM_at - -
          Murine Genome U74v2 Set 102971_at - -
          Mouse Genome 430A 2.0 Array 1422105_at - -
          Mouse Genome 430A 2.0 Array 1422105_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422105_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422105_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3800056 - -
          Illumina mouse-6 v1.0 expression beadchip 3800056 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003800056 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1238847 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1238847 - -
          mouse-6 v1.1 (Illumina) scl012501.1_271-S - -
          Illumina Sentrix mouse-6.v1 bead array 3800056 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl012501.1_271-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3800056 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2784580 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1238847 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2784580 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline