Search
Search

Kmt2a| Gene Symbol: Kmt2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081049.1 | NP_001074518.1 | 9 - 10 | 4213 | 66 | |
| GenBank mRNA | AK140439.1 | - | 7 - 8 | 2956 | 66 | |
| L17069.1 | - | 9 - 10 | 3915 | 66 | ||
Set Membership: |
|