Search
Search

Adprhl2| Gene Symbol: Adprhl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133883.2 | NP_598644.1 | 1 - 2 | 261 | 102 | |
| GenBank mRNA | AJ427296.1 | - | 1 - 2 | 261 | 102 | |
| AK143583.1 | - | 1 - 2 | 251 | 102 | ||
| AK147034.1 | - | 1 - 2 | 237 | 102 | ||
| AK149904.1 | - | 1 - 2 | 253 | 102 | ||
| AK150202.1 | - | 1 - 2 | 253 | 102 | ||
| AK169070.1 | - | 1 - 2 | 248 | 102 | ||
| AK171780.1 | - | 1 - 2 | 251 | 102 | ||
Set Membership: |
|