Search
Search

Zmym1| Gene Symbol: Zmym1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026670.4 | NP_080946.3 | 7 - 8 | 1020 | 80 | |
| GenBank mRNA | AK016783.1 | - | 7 - 8 | 1016 | 80 | |
| AK035618.1 | - | 7 - 8 | 1020 | 80 | ||
| AK049568.1 | - | 7 - 8 | 1010 | 80 | ||
| AK132399.1 | - | 3 - 4 | 1982 | 80 | ||
| AK166769.1 | - | 7 - 8 | 1003 | 80 | ||
| AK167508.1 | - | 7 - 8 | 1026 | 80 | ||
| BC043454.1 | - | 6 - 7 | 637 | 80 | ||
| BC057203.1 | - | 4 - 5 | 434 | 80 | ||
Set Membership: |
|