Search
Search

Tfap2e| Gene Symbol: Tfap2e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198960.2 | NP_945198.2 | 3 - 4 | 841 | 55 | |
| GenBank mRNA | AY325902.1 | - | 3 - 4 | 681 | 55 | |
| BI134735.1 | - | 2 - 3 | 251 | 55 | ||
Set Membership: |
|