Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01180221_g1
          See other GAPDH GE Assays ›
          Gene Symbol:
          Gapdh Gm12070 Gm5523 Gm6981
          Gene Name:
          glyceraldehyde-3-phosphate dehydrogenase
          glyceraldehyde-3-phosphate dehydrogenase pseudogene
          glyceraldehyde-3-phosphate dehydrogenase pseudogene
          glyceraldehyde-3-phosphate dehydrogenase pseudogene
          Gene Aliases:
          Gapd
          OTTMUSG00000005300
          EG433273
          EG629557
          Chromosome Location:
          Chr.6: 125161338 - 125166511 on Build GRCm38
          Chromosome Location:
          Chr.11: 26785108 - 26787860 on Build GRCm38
          Chromosome Location:
          Chr.1: 13785685 - 13786929 on Build GRCm38
          Chromosome Location:
          Chr.9: 52002061 - 52048705 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          Assay ID Mm01180221_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gapdh
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001289726.1 NP_001276655.1 4 527 80
          NM_008084.3 NP_032110.1 4 675 80
          GenBank mRNA AK002273.1 - 4 508 80
          AK081405.1 - 4 508 80
          AK140794.1 - 4 508 80
          AK144690.1 - 4 529 80
          AK146435.1 - 4 508 80
          AK147738.1 - 4 508 80
          AK147891.1 - 4 508 80
          AK160399.1 - 4 508 80
          AK164415.1 - 4 506 80
          AK168217.1 - 4 508 80
          AK169742.1 - 1 2078 80
          AK178680.1 - 1 76 80
          AK184664.1 - 1 77 80
          AK184690.1 - 1 85 80
          AK197268.1 - 1 76 80
          AK200636.1 - 1 76 80
          AK201486.1 - 1 76 80
          AK204478.1 - 1 77 80
          BC020407.1 - 4 496 80
          BC023196.1 - 4 506 80
          BC082592.1 - 4 485 30634381 80
          BC083065.1 - 4 484 5371133 80
          BC083079.1 - 4 490 6493407 80
          BC083080.1 - 4 492 6494145 80
          BC083149.1 - 471 30459585 80
          BC085274.1 - 493 6530979 80
          BC085275.1 - 4 502 30444086 80
          BC091768.1 - 471 4159824 80
          BC092252.1 - 4 494 5044783 80
          BC092264.1 - 496 2655526 80
          BC092267.1 - 482 4010884 80
          BC092294.1 - 485 5068049 80
          BC093508.1 - 465 4975394 80
          BC094037.1 - 472 5009636 80
          BC095932.1 - 4 496 30733532 80
          BC096440.1 - 4 496 2649847 80
          BC096590.1 - 496 6486174 80
          BC110311.1 - 4 497 5340298 80
          BC145810.1 - 4 478 40131113 80
          BC145812.1 - 4 478 40131115 80
          BU504528.1 - 493 80
          DQ403054.1 - 2 366 80
          GU214026.1 - 4 506 80
          M32599.1 - 4 481 80
          Gene Symbol: Gm12070
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_002890.1 2 854 80
          GenBank mRNA BC098095.1 - 2 854 80
          Gene Symbol: Gm5523
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_004447.1 1 480 80
          GenBank mRNA AK188672.1 - 85 80
          AK193983.1 - 76 80
          Gene Symbol: Gm6981
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_023357.1 3 80
          GenBank mRNA AK132984.1 - 3 80

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Both primers and probe map within a single exon

          Panther Classification:

          Molecular Function -

          dehydrogenase

          Gene Ontology Categories:

          Function(s) Process(es)

          microtubule cytoskeleton organization
          carbohydrate metabolic process
          gluconeogenesis
          glycolytic process
          apoptotic process
          multicellular organism development
          negative regulation of translation
          peptidyl-cysteine S-trans-nitrosylation
          protein stabilization
          neuron apoptotic process
          oxidation-reduction process
          response to ammonium ion
          cellular response to interferon-gamma
          glyceraldehyde-3-phosphate dehydrogenase (NAD+) (phosphorylating) activity
          microtubule binding
          peptidyl-cysteine S-nitrosylase activity
          identical protein binding

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2106956 NR_004447.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2034600 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2034600 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10455781 - -
          Affymetrix Mouse Gene 1.0 ST Array 10353191 - -
          Affymetrix Mouse Gene 1.0 ST Array 10426436 - -
          Affymetrix Mouse Gene 1.0 ST Array 10492659 - -
          Affymetrix Mouse Gene 1.0 ST Array 10463065 - -
          Affymetrix Mouse Gene 1.0 ST Array 10353190 - -
          Affymetrix Mouse Gene 1.0 ST Array 10601568 - -
          Affymetrix Mouse Gene 1.1 ST Array 10353189 - -
          Affymetrix Mouse Clariom S Array PSR0100016232.mm.1 NR_004447.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100016232.mm.1 TC0100001983.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100016233.mm.1 NR_004447.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100016233.mm.1 TC0100001983.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100016231.mm.1 NR_004447.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100016231.mm.1 TC0100001983.mm.1 MTA TC
          Mouse Expression Set 430 1418625_PM_s_at - -
          Mouse Expression Set 430 1418625_s_at - -
          Mouse Genome 430A 2.0 Array 1418625_s_at - -
          Mouse Genome 430A 2.0 Array 1418625_PM_s_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline