Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01180531_m1
          See other HDDC2 GE Assays ›
          Gene Symbol:
          Hddc2
          Gene Name:
          HD domain containing 2
          Gene Aliases:
          2310057G13Rik
          Chromosome Location:
          Chr.10: 31313362 - 31328086 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01180531_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hddc2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027168.2 NP_081444.1 2 - 3 206 70
          GenBank mRNA AK009957.1 - 2 - 3 147 70
          AK161186.1 - 1 - 2 1237 70
          BC104360.1 - 2 - 3 206 40049707 70
          BC104361.1 - 2 - 3 206 40049709 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000000279.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hddc2

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27043768 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27043770 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2004930 NM_027168.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2004930 AK009957.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2004930 BC104360.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2004930 BC104361.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 96971 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 371814 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 868214 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 914272 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 254977 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 779621 AK008052.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 779621 AK136525.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 779621 AK153247.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 779621 BC003493.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 779621 NM_053217.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 804245 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 964785 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 112871 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 558942 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 76774 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 768 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P160544 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P117927 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2004930 NM_027168.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2004930 AK009957.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2004930 BC104360.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2004930 BC104361.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27043744 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27043756 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27043755 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27043762 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27043751 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27043789 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27043782 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27043776 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27043770 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27043749 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27043764 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27043746 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27043792 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27043743 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27043758 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27043779 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27043791 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27043765 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27043766 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27043786 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27043750 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27043754 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27043763 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27043774 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27043785 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27043771 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27043775 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27043769 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27043781 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27043748 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27043783 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P160544 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02096251 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P117927 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07019021 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P160544 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P117927 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27043768 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27043770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27043779 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043812 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043733 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043765 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043824 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08712333 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043756 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043879 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043803 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043800 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043738 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043785 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043737 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043774 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043818 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043771 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043751 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043724 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043823 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043741 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043783 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08712336 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043822 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043793 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043748 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043883 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043852 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08696423 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043796 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043870 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043814 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043746 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043725 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043804 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043744 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08808986 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043786 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08231087 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043775 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08727053 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043808 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043847 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043886 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043740 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08517742 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043854 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043770 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043874 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043889 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043873 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043761 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043739 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043779 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043735 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043851 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043767 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043850 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08808989 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043762 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043815 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043750 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043810 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08909110 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08259961 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043789 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043763 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043875 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08428784 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043749 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043731 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043754 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043817 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043869 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043878 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043794 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043782 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043848 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043766 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08259956 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043791 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043871 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27043820 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10362399 - -
          Affymetrix Mouse Gene 1.0 ST Array 10362400 - -
          Affymetrix Mouse Gene 1.0 ST Array 10362396 - -
          Affymetrix Mouse Gene 1.0 ST Array 10362398 - -
          Affymetrix Mouse Gene 1.0 ST Array 10362397 - -
          Affymetrix Mouse Gene 1.0 ST Array 10362395 - -
          Affymetrix Mouse Gene 1.0 ST Array 10362401 - -
          Affymetrix HT MG-430 PM Array Plate 1428823_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10362394 - -
          Mouse Genome 430 2.0 Array 1428823_at - -
          Mouse Genome 430 2.0 Array 1428823_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69692_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69692_at - -
          Affymetrix Mouse Clariom S Array PSR1000001872.mm.1 NM_027168.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000001872.mm.1 NM_027168.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000001872.mm.1 TC1000000279.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000001868.mm.1 NM_027168.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000001868.mm.1 NM_027168.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000001868.mm.1 TC1000000279.mm.1 MTA TC
          Mouse Expression Set 430 1428823_PM_at - -
          Mouse Expression Set 430 1428823_at - -
          Murine Genome U74v2 Set 160659_PM_at - -
          Murine Genome U74v2 Set 160659_at - -
          Mouse Genome 430A 2.0 Array 1428823_PM_at - -
          Mouse Genome 430A 2.0 Array 1428823_at - -
          Affymetrix HT Mouse Genome 430A Array 1428823_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1428823_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 830184 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1244187 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2638781 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2638781 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1244187 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0