Search
Search

Zmym4| Gene Symbol: Zmym4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114399.1 | NP_001107871.1 | 16 - 17 | 2805 | 62 | |
| GenBank mRNA | AK129137.1 | - | 16 - 17 | 3060 | 62 | |
| BC050924.1 | - | 10 - 11 | 1714 | 62 | ||
Set Membership: |
|