| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1000002595.mm.1 |
NM_017472.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000002595.mm.1 |
NM_017472.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000002595.mm.1 |
TC1000000382.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000002598.mm.1 |
NM_017472.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000002598.mm.1 |
NM_017472.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000002598.mm.1 |
TC1000000382.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000002594.mm.1 |
NM_017472.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000002594.mm.1 |
NM_017472.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000002594.mm.1 |
TC1000000382.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000002593.mm.1 |
NM_017472.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000002593.mm.1 |
NM_017472.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000002593.mm.1 |
TC1000000382.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000002596.mm.1 |
NM_017472.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000002596.mm.1 |
NM_017472.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000002596.mm.1 |
TC1000000382.mm.1 |
MTA TC |