Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01181863_m1
          See other 2010001E11RIK GE Assays ›
          Gene Symbol:
          2010001E11Rik
          Gene Name:
          RIKEN cDNA 2010001E11 gene
          Gene Aliases:
          Naglt1b
          Chromosome Location:
          Chr.10: 39920382 - 39926923 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01181863_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 2010001E11Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001163503.1 NP_001156975.1 2 - 3 357 132
          GenBank mRNA AK008003.1 - 2 - 3 358 132
          BC145051.1 - 525 132

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000002031.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          ion transport
          sodium ion transport
          biological_process
          carbohydrate transport
          transmembrane transport
          molecular_function
          symporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1994812 NM_001163503.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1994812 AK008003.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 666026 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 439096 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 32134 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 865770 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 14086 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 39269 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 124281 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P141665 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1994812 NM_001163503.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1994812 AK008003.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P141665 - -
          Whole Mouse Genome G4122A A_51_P505846 NM_001163503.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P505846 AK008003.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P505846 BC145051.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P141665 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27081432 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27081456 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27081439 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27081421 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27081458 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27081397 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27081424 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27081389 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27081379 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27081403 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27081376 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08750458 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27081449 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27081437 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27081462 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27081453 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27081383 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08668414 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27081433 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27081387 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27081450 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27081419 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27081399 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27081436 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27081388 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27081451 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27081406 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08668407 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27081398 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27081461 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08317121 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27081441 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27081430 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27081447 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27081443 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27081431 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27081381 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08510439 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27081422 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10368696 - -
          Affymetrix Mouse Gene 1.0 ST Array 10368698 - -
          Affymetrix Mouse Gene 1.0 ST Array 10368697 - -
          Affymetrix HT MG-430 PM Array Plate 1431537_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10368695 - -
          Mouse Genome 430 2.0 Array 1431537_PM_at - -
          Mouse Genome 430 2.0 Array 1431537_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 72045_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 72045_at - -
          Affymetrix Mouse Clariom S Array PSR1000014426.mm.1 NM_001163503.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000014426.mm.1 NM_001163503.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000014426.mm.1 TC1000002031.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000014427.mm.1 NM_001163503.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000014427.mm.1 NM_001163503.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000014427.mm.1 TC1000002031.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000014425.mm.1 NM_001163503.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000014425.mm.1 NM_001163503.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000014425.mm.1 TC1000002031.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000014424.mm.1 NM_001163503.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000014424.mm.1 NM_001163503.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000014424.mm.1 TC1000002031.mm.1 MTA TC
          Mouse Expression Set 430 1451760_s_at - -
          Mouse Expression Set 430 1451760_PM_s_at - -
          Mouse Expression Set 430 1431537_PM_at - -
          Mouse Expression Set 430 1431537_at - -
          Mouse Genome 430A 2.0 Array 1451760_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1451760_s_at - -
          Affymetrix HT Mouse Genome 430B Array 1431537_at - -
          Affymetrix HT Mouse Genome 430B Array 1431537_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 470713 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline