Search
Search

Magi1| Gene Symbol: Magi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029850.4 | NP_001025021.1 | 12 - 13 | 2741 | 72 | |
| NM_001083320.2 | NP_001076789.1 | 11 - 12 | 2083 | 72 | ||
| NM_001083321.2 | NP_001076790.1 | 11 - 12 | 2083 | 72 | ||
| NM_001286784.1 | NP_001273713.1 | 11 - 12 | 2705 | 72 | ||
| NM_001286785.1 | NP_001273714.1 | 12 - 13 | 2741 | 72 | ||
| NM_001286786.1 | NP_001273715.1 | 11 - 12 | 2705 | 72 | ||
| NM_001286788.1 | NP_001273717.1 | 11 - 12 | 2083 | 72 | ||
| NM_010367.3 | NP_034497.1 | 11 - 12 | 2705 | 72 | ||
| GenBank mRNA | AB194411.1 | - | 11 - 12 | 2110 | 72 | |
| AB194412.1 | - | 11 - 12 | 2110 | 72 | ||
| AF027503.1 | - | 11 - 12 | 3034 | 72 | ||
| AK147358.1 | - | 12 - 13 | 2742 | 72 | ||
| AK163582.1 | - | 8 - 9 | 1408 | 72 | ||
| AK163800.1 | - | 11 - 12 | 1948 | 72 | ||
| AK220499.1 | - | 11 - 12 | 1788 | 72 | ||
| AY497557.1 | - | 12 - 13 | 2146 | 72 | ||
| BC060273.1 | - | 11 - 12 | 2083 | 72 | ||
| BC095943.1 | - | 12 - 13 | 2719 | 72 | ||
| BC150820.1 | - | 12 - 13 | 2415 | 9087731 | 72 | |
Set Membership: |
|