Search
Search
| Gene Symbol: Cd8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081110.2 | NP_001074579.1 | 2 - 3 | 675 | 68 | |
| NM_009857.1 | NP_033987.1 | 2 - 3 | 675 | 68 | ||
| GenBank mRNA | AK037467.1 | - | 2 - 3 | 554 | 68 | |
| AK088128.1 | - | 2 - 3 | 555 | 68 | ||
| AK088214.1 | - | 2 - 3 | 555 | 68 | ||
| AK088414.1 | - | 2 - 3 | 550 | 68 | ||
| AK088447.1 | - | 1 - 2 | 1060 | 68 | ||
| AK137937.1 | - | 2 - 3 | 526 | 68 | ||
| BC030679.1 | - | 2 - 3 | 539 | 1247019 | 68 | |
| M12052.1 | - | 2 - 3 | 675 | 68 | ||
| M12825.1 | - | 2 - 3 | 561 | 68 | ||
| M16981.1 | - | 2 - 3 | 558 | 68 | ||
| U34881.1 | - | 2 - 3 | 549 | 68 | ||
Set Membership: |
|