Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01182346_m1
          See other 1700049E22RIK GE Assays ›
          Gene Symbol:
          1700049E22Rik
          Gene Name:
          RIKEN cDNA 1700049E22 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 100527400 - 100533426 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01182346_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700049E22Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040525.1 2 - 3 321 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600001152.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Recommended Assay Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2172713 AK006738.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 697499 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 138246 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P105967 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2172713 AK006738.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P105967 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P136391 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P105967 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24692647 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24692565 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05463592 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05787111 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05217271 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05868956 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05266211 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24692680 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24692623 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05727361 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05136065 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05510717 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24692617 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05427841 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24692674 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24692587 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24692667 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05217272 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24692625 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05463594 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05136066 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24692703 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05266212 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24692662 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05748655 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24692634 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05158039 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24692566 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05217277 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24692609 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05217268 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24692659 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05868949 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24692584 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24692675 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05845742 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05463227 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05855220 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24692591 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05463589 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24692687 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24692651 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24692638 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05409510 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05812073 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05855218 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05868969 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24692700 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24692657 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05594694 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24692682 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24692673 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24692702 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24692588 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05467075 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24692640 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05855222 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24692686 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24692602 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24692676 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24692614 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05868950 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05845740 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24692641 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24692639 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24692699 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24692616 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05855219 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05803126 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24692589 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24692637 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05136067 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24692571 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24692695 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05868967 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05812069 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24692619 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0600008919.mm.1 TC0600001152.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600008919.mm.1 NR_040525.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600008919.mm.1 NR_040525.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600008920.mm.1 TC0600001152.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600008920.mm.1 NR_040525.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600008920.mm.1 NR_040525.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600008917.mm.1 TC0600001152.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600008917.mm.1 NR_040525.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600008917.mm.1 NR_040525.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 395950 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline