| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P427964 |
NM_001081157.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P427964 |
AK030676.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P427964 |
NM_001081157.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P427964 |
AK030676.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
897413 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
517758 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
343593 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
114330 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
940889 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
635401 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
931864 |
AK008050.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
931864 |
AK008140.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
931864 |
AK008470.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
931864 |
D63356.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
931864 |
D63357.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
931864 |
NM_011259.1 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
283435 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
825089 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
77768 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P167946 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P427964 |
NM_001081157.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P427964 |
AK030676.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P427964 |
NM_001081157.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P427964 |
AK030676.1 |
mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P01394199 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P167946 |
- |
- |
| Whole Mouse Genome G4122A |
A_51_P427964 |
NM_001081157.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P427964 |
AK030676.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P427964 |
NM_001081157.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P427964 |
AK030676.1 |
mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P167946 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24672049 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24671982 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24671949 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24671939 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24671933 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24672035 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24671950 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24672019 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24671883 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24671962 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24671987 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24671925 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24672047 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24672045 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24671969 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24671941 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24672023 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24672039 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24672078 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24671984 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24672012 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24672009 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24671934 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24671993 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24671968 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24671895 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24671957 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24671881 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24672051 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24671893 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24672067 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24672062 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24671990 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24672015 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24671937 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24671885 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24671880 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24671882 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24672013 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24672014 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24672050 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24671947 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24672077 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24672003 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24672017 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24672044 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24672024 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24672028 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24671992 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24671995 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24672018 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24671891 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24671896 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24671887 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24672048 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24671980 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24671997 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24672005 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24671975 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24671985 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24671967 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24672021 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24671960 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24671951 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24672030 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24671976 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24671943 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24672032 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24671988 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24671999 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24672073 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24672065 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24671991 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24672027 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24671973 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24672034 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24671894 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24671952 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24672000 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24672055 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24672053 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24671988 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P05427966 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24671985 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P05502677 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P05382805 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24672023 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24672065 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P05630217 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24671993 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P05271178 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24672017 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24671976 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24672015 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P05568851 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24671943 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P05171117 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24672001 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24672012 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24672009 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24672069 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24671950 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24671982 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24671947 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24671960 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24671971 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24672014 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24672004 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24671968 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24672045 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24671945 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24672035 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24672000 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P05427964 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24671987 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24672018 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P05882085 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24671997 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P05849337 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24671957 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P05382795 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24671999 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24672075 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P05464566 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24672051 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24672024 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P05427959 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24672044 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24672042 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24671969 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24672027 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24671995 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P05271180 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24672010 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24672053 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24672049 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P05370325 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24672047 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24672067 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24672048 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24671967 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24671975 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24672073 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24671949 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P05038981 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P05032709 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24672003 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P05492583 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24672019 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24672050 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24672005 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P05382803 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P05403331 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24672080 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24672016 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P05099091 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24672037 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24672013 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P24672041 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P05706273 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P05427971 |
- |
- |