Search
Search

Tmem52b| Gene Symbol: Tmem52b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081186.1 | NP_001074655.1 | 2 - 3 | 589 | 62 | |
| GenBank mRNA | AK085571.1 | - | 2 - 3 | 529 | 62 | |
| BC120661.1 | - | 2 - 3 | 242 | 40129584 | 62 | |
| BC120663.1 | - | 2 - 3 | 242 | 40129586 | 62 | |
Set Membership: |
|