Search
Search

Bmp10| Gene Symbol: Bmp10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009756.2 | NP_033886.2 | 1 - 2 | 362 | 74 | |
| GenBank mRNA | BC145044.1 | - | 1 - 2 | 362 | 9053592 | 74 |
| BC145983.1 | - | 1 - 2 | 362 | 40131286 | 74 | |
Set Membership: |
|