Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01184198_g1
          See other DPPA3 GE Assays ›
          Gene Symbol:
          Dppa3
          Gene Name:
          developmental pluripotency-associated 3
          Gene Aliases:
          2410075G02Rik, PCG7, PGC7, Stella
          Chromosome Location:
          Chr.6: 122626424 - 122630271 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01184198_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dppa3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_139218.1 NP_631964.1 3 - 4 454 89
          GenBank mRNA AB072734.1 - 3 - 4 355 89
          AF490347.1 - 3 - 4 459 89
          AK010723.1 - 3 - 4 395 89
          AK135769.1 - 3 - 4 459 89
          AK136263.1 - 3 - 4 459 89
          AY082485.1 - 3 - 4 454 89
          AY134859.1 - 3 - 4 466 89
          BC099433.1 - 3 - 4 432 30933569 89
          BC100331.1 - 3 - 4 429 30934920 89
          BC107340.1 - 3 - 4 386 40055218 89
          BC107341.1 - 3 - 4 386 40055220 89

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dppa3

          Gene Ontology Categories:

          Function(s) Process(es)

          multicellular organism development
          chromatin modification
          embryonic cleavage
          protection of DNA demethylation of female pronucleus
          negative regulation of DNA demethylation
          regulation of genetic imprinting
          protein binding
          methylated histone binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2091496 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 707003 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 240753 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 655684 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P169338 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P153601 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P160435 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2091496 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24809624 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24809621 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24809631 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24809635 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24809619 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24809640 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24809646 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24809626 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24809617 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24809628 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24809636 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24809633 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24809639 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24809654 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24809657 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24809651 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24809637 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24809649 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24809623 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24809645 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P169338 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P160435 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P153601 - -
          Whole Mouse Genome G4122A A_51_P195044 AK136263.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P195044 AY082485.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P195044 AK135769.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P195044 AY134859.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P195044 BC099433.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129420 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P153601 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P160435 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P169338 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05765875 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05422379 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05765874 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05422381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809696 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24809677 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24809639 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05380027 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24809633 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05252791 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24809682 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24809661 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05017239 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24809637 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24809636 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24809628 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24809646 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24809674 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24809651 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24809681 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05723303 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24809668 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05422381 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24809621 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24809671 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24809679 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24809629 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05723302 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24809638 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24809673 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05298620 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24809624 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05017234 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24809672 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24809627 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24809663 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24809684 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24809640 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24809631 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05422378 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24809666 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24809654 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05422379 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24809657 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24809688 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24809680 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05865219 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05141793 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24809635 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10604138 - -
          Affymetrix Mouse Gene 1.0 ST Array 10604137 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541518 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541516 - -
          Affymetrix Mouse Gene 1.0 ST Array 10604136 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541519 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541517 - -
          Affymetrix HT MG-430 PM Array Plate 1424295_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10541515 - -
          Affymetrix Mouse Gene 1.1 ST Array 10604135 - -
          Mouse Genome 430 2.0 Array 1424295_at - -
          Mouse Genome 430 2.0 Array 1424295_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 73708_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 73708_at - -
          Affymetrix Mouse Clariom S Array PSR0600011195.mm.1 TC0600001388.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600011195.mm.1 NM_139218.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011195.mm.1 NM_139218.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011193.mm.1 TC0600001388.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600011193.mm.1 NM_139218.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011193.mm.1 NM_139218.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011194.mm.1 TC0600001388.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600011194.mm.1 NM_139218.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011194.mm.1 NM_139218.1 Refseq
          Mouse Expression Set 430 1424295_PM_at - -
          Mouse Expression Set 430 1424295_at - -
          Murine Genome U74v2 Set 97283_PM_at - -
          Murine Genome U74v2 Set 97283_at - -
          Mouse Genome 430A 2.0 Array 1424295_PM_at - -
          Mouse Genome 430A 2.0 Array 1424295_at - -
          Affymetrix HT Mouse Genome 430A Array 1424295_at - -
          Affymetrix HT Mouse Genome 430A Array 1424295_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1410341 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001410341 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1225176 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1225176 - -
          mouse-6 v1.1 (Illumina) scl073708.2_25-S - -
          Illumina Sentrix mouse-6.v1 bead array 1410341 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl073708.2_25-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1410341 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1225176 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1225176 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline