| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0600010115.mm.1 |
TC0600001281.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600010115.mm.1 |
NM_199033.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600010115.mm.1 |
NM_199033.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600010114.mm.1 |
TC0600001281.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600010114.mm.1 |
NM_199033.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600010114.mm.1 |
NM_199033.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600010112.mm.1 |
TC0600001281.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600010112.mm.1 |
NM_199033.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600010112.mm.1 |
NM_199033.1 |
Refseq |