| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0600012310.mm.1 |
TC0600001509.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600012310.mm.1 |
NM_001162858.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600012310.mm.1 |
NM_001162858.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600012310.mm.1 |
NM_021717.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600012310.mm.1 |
NM_021717.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600012313.mm.1 |
TC0600001509.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600012313.mm.1 |
NM_001162858.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600012313.mm.1 |
NM_001162858.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600012313.mm.1 |
NM_021717.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600012313.mm.1 |
NM_021717.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600012316.mm.1 |
TC0600001509.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600012316.mm.1 |
NM_001162858.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600012316.mm.1 |
NM_001162858.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600012316.mm.1 |
NM_021717.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600012316.mm.1 |
NM_021717.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600012312.mm.1 |
TC0600001509.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600012312.mm.1 |
NM_001162858.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600012312.mm.1 |
NM_001162858.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600012312.mm.1 |
NM_021717.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600012312.mm.1 |
NM_021717.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600012314.mm.1 |
TC0600001509.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600012314.mm.1 |
NM_001162858.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600012314.mm.1 |
NM_001162858.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600012314.mm.1 |
NM_021717.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600012314.mm.1 |
NM_021717.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600012318.mm.1 |
TC0600001509.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600012318.mm.1 |
NM_001162858.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600012318.mm.1 |
NM_001162858.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600012318.mm.1 |
NM_021717.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600012318.mm.1 |
NM_021717.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600012311.mm.1 |
TC0600001509.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600012311.mm.1 |
NM_001162858.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600012311.mm.1 |
NM_001162858.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600012311.mm.1 |
NM_021717.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600012311.mm.1 |
NM_021717.2 |
Refseq |