Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01184749_m1
          See other FXYD4 GE Assays ›
          Gene Symbol:
          Fxyd4
          Gene Name:
          FXYD domain-containing ion transport regulator 4
          Gene Aliases:
          0610008I02Rik, AI267073, Chif
          Chromosome Location:
          Chr.6: 117933559 - 117938510 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01184749_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fxyd4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001173372.1 NP_001166843.1 3 - 4 157 68
          NM_033648.1 NP_387468.1 3 - 4 160 68
          GenBank mRNA AK018728.1 - 3 - 4 157 68
          AK054004.1 - 2 - 3 976 68
          BC086918.1 - 4 - 5 421 6762406 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600002992.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fxyd4

          Panther Classification:

          Molecular Function -

          primary active transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          ion transport
          potassium ion transmembrane transport
          regulation of sodium ion transmembrane transporter activity
          ion channel activity
          potassium channel activity
          sodium channel regulator activity
          ATPase binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P141926 AK018728.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P141926 AU041511.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P141926 BC086918.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P141926 NM_001173372.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P141926 NM_033648.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P141926 AK018728.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P141926 AU041511.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P141926 BC086918.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P141926 NM_001173372.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P141926 NM_033648.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 520530 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 80622 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P136212 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P141926 AK018728.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P141926 AU041511.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P141926 BC086918.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P141926 NM_001173372.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P141926 NM_033648.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P141926 AK018728.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P141926 AU041511.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P141926 BC086918.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P141926 NM_001173372.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P141926 NM_033648.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P141926 AK018728.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P141926 AU041511.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P141926 BC086918.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P141926 NM_001173372.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P141926 NM_033648.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P141926 AK018728.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P141926 AU041511.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P141926 BC086918.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P141926 NM_001173372.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P141926 NM_033648.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24784155 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24784154 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24784167 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24784157 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24784144 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24784152 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24784134 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24784165 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24784150 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24784143 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24784141 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24784164 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24784158 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24784128 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24784137 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24784135 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24784140 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24784130 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24784151 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24784160 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24784146 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24784161 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24784148 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24784131 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24784138 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24784169 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P136212 - -
          Whole Mouse Genome G4122A A_51_P141926 AK018728.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P141926 AU041511.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P141926 BC086918.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P141926 NM_001173372.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P141926 NM_033648.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P141926 AK018728.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P141926 AU041511.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P141926 BC086918.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P141926 NM_001173372.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P141926 NM_033648.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112581 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P136212 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05601541 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05617217 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05617218 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05617222 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05601540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24784160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24784150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24784167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24784116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24784143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24784114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24784102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24784119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24784146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24784111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24784100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24784124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24784130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24784120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24784154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24784152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24784106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24784161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24784148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24784175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24784128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24784135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24784125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24784107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24784164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24784108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24784137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24784134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24784165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24784172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24784170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24784157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24784103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24784138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24784131 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24784143 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24784103 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05439486 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24784144 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24784141 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24784170 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24784142 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24784172 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24784149 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24784108 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05617224 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24784131 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24784150 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24784160 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05104075 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24784128 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24784164 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24784118 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24784161 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24784134 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05104082 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24784165 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05733725 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05326894 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24784173 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05046160 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24784175 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24784097 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24784098 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05372422 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05617221 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24784156 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05706888 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05349621 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24784114 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24784119 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24784102 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24784137 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24784111 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24784116 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24784120 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24784152 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05799561 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24784167 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05570415 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24784155 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24784138 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24784157 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24784132 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05349626 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24784110 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24784106 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24784107 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05777265 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10547208 - -
          Affymetrix Mouse Gene 1.0 ST Array 10547212 - -
          Affymetrix Mouse Gene 1.0 ST Array 10547213 - -
          Affymetrix Mouse Gene 1.0 ST Array 10547209 - -
          Affymetrix Mouse Gene 1.0 ST Array 10547207 - -
          Affymetrix Mouse Gene 1.0 ST Array 10547214 - -
          Affymetrix Mouse Gene 1.0 ST Array 10547216 - -
          Affymetrix Mouse Gene 1.0 ST Array 10547215 - -
          Affymetrix Mouse Gene 1.0 ST Array 10547210 - -
          Affymetrix Mouse Gene 1.0 ST Array 10547211 - -
          Affymetrix HT MG-430 PM Array Plate 1418207_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10547206 - -
          Mouse Genome 430 2.0 Array 1418207_at - -
          Mouse Genome 430 2.0 Array 1418207_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 108017_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 108017_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0600022990.mm.1 TC0600002992.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600022990.mm.1 NM_001173372.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600022990.mm.1 NM_001173372.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600022990.mm.1 NM_033648.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600022990.mm.1 NM_033648.1 Refseq
          Mouse Expression Set 430 1418207_PM_at - -
          Mouse Expression Set 430 1418207_at - -
          Murine Genome U74v2 Set 171114_at - -
          Murine Genome U74v2 Set 171114_PM_at - -
          Mouse Genome 430A 2.0 Array 1418207_at - -
          Mouse Genome 430A 2.0 Array 1418207_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1418207_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1418207_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 704669 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5270292 - -
          Illumina mouse-6 v1.0 expression beadchip 5270292 - -
          Illumina mouse-6 v1.0 expression beadchip 6370725 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005270292 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006370725 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2670751 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2720117 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2720117 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2670751 - -
          mouse-6 v1.1 (Illumina) scl0001091.1_5-S - -
          mouse-6 v1.1 (Illumina) scl28501.8.1_119-S - -
          Illumina Sentrix mouse-6.v1 bead array 5270292 - -
          Illumina Sentrix mouse-6.v1 bead array 6370725 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0001091.1_5-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl28501.8.1_119-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6370725 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5270292 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1215552 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2670751 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1215552 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2670751 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline