Search
Search

Ankrd26| Gene Symbol: Ankrd26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081112.1 | NP_001074581.1 | 22 - 23 | 2667 | 65 | |
| GenBank mRNA | AK173096.1 | - | 6 - 7 | 856 | 65 | |
| BC008111.1 | - | 2 - 3 | 213 | 65 | ||
| BC056364.1 | - | 5 - 6 | 726 | 65 | ||
Set Membership: |
|