Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01184860_g1
          See other DOK1 GE Assays ›
          Gene Symbol:
          Dok1
          Gene Name:
          docking protein 1
          Gene Aliases:
          AW557123, p62DOK
          Chromosome Location:
          Chr.6: 83030932 - 83033471 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01184860_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dok1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001291799.1 NP_001278728.1 3 - 4 494 70
          NM_010070.4 NP_034200.4 3 - 4 496 70
          GenBank mRNA AK136382.1 - 3 - 4 497 70
          AK169871.1 - 3 - 4 492 70
          BC013066.1 - 3 - 4 490 3498170 70
          CJ053718.1 - 3 - 4 497 70
          U78818.2 - 3 - 4 461 70

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dok1

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          transmembrane receptor protein tyrosine kinase signaling pathway
          Ras protein signal transduction
          intracellular signal transduction
          receptor signaling protein activity
          insulin receptor binding
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24590979 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24590984 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24590977 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24590980 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24590981 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P275976 BC013066.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P275976 BI135039.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P275976 U78818.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P275976 NM_010070.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P275976 AK136382.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P275976 BC013066.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P275976 BI135039.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P275976 U78818.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P275976 NM_010070.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P275976 AK136382.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 941867 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 879697 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 89594 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P275976 BC013066.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P275976 BI135039.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P275976 U78818.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P275976 NM_010070.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P275976 AK136382.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P275976 BC013066.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P275976 BI135039.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P275976 U78818.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P275976 NM_010070.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P275976 AK136382.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P275976 BC013066.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P275976 BI135039.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P275976 U78818.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P275976 NM_010070.4 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P275976 AK136382.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P275976 BC013066.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P275976 BI135039.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P275976 U78818.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P275976 NM_010070.4 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P275976 AK136382.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P366842 BC013066.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P366842 BI135039.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P366842 U78818.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P366842 NM_010070.4 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P366842 AK136382.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24590961 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24590984 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24590982 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24590959 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24590963 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24590976 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24590980 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24590971 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24590988 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24590958 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24590964 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24590966 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24590965 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24590977 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24590985 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24590960 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24590967 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24590973 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24590969 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24590957 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05960418 - -
          Whole Mouse Genome G4122A A_52_P366842 BC013066.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P366842 BI135039.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P366842 U78818.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P366842 NM_010070.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P366842 AK136382.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P275976 BC013066.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P275976 BI135039.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P275976 U78818.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P275976 NM_010070.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P275976 AK136382.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P275976 BC013066.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P275976 BI135039.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P275976 U78818.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P275976 NM_010070.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P275976 AK136382.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P139265 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24590981 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24590984 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24590980 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24590977 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24590979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24590977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24590985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24590982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24590980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24590971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24590966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24590973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24590963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24590969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24590965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24590958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24590957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24590967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24590976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24590988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24590960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24590984 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24590963 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24590965 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24590980 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24590973 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24590957 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24590971 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24590967 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24590966 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24590959 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24590969 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24590958 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24590960 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24590982 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24590985 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24590976 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24590977 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10545627 - -
          Affymetrix Mouse Gene 1.0 ST Array 10545624 - -
          Affymetrix Mouse Gene 1.0 ST Array 10545625 - -
          Affymetrix Mouse Gene 1.0 ST Array 10545626 - -
          Affymetrix Mouse Gene 1.0 ST Array 10545628 - -
          Affymetrix HT MG-430 PM Array Plate 1417790_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10545623 - -
          Mouse Genome 430 2.0 Array 1417790_PM_at - -
          Mouse Genome 430 2.0 Array 1417790_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 13448_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 13448_at - -
          Affymetrix Mouse Clariom S Array PSR0600019771.mm.1 TC0600002554.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600019771.mm.1 NM_010070.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600019771.mm.1 NM_010070.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0600019770.mm.1 TC0600002554.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600019770.mm.1 NM_010070.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600019770.mm.1 NM_010070.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0600019768.mm.1 TC0600002554.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600019768.mm.1 NM_010070.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600019768.mm.1 NM_010070.4 Refseq
          Mouse Expression Set 430 1417790_at - -
          Mouse Expression Set 430 1417790_PM_at - -
          Murine Genome U74v2 Set 102896_PM_at - -
          Murine Genome U74v2 Set 102896_at - -
          Mouse Genome 430A 2.0 Array 1417790_PM_at - -
          Mouse Genome 430A 2.0 Array 1417790_at - -
          Affymetrix HT Mouse Genome 430A Array 1417790_at - -
          Affymetrix HT Mouse Genome 430A Array 1417790_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 657562 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5220273 - -
          MouseWG-6 v2 Expression BeadChip 2680112 - -
          MouseWG-6 v2 Expression BeadChip 1050279 - -
          Illumina mouse-6 v1.0 expression beadchip 5220273 - -
          Illumina mouse-6 v1.0 expression beadchip 2680112 - -
          Illumina mouse-6 v1.0 expression beadchip 1050279 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002680112 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005220273 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001050279 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2704141 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1242767 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1257069 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1242767 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2704141 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1257069 - -
          mouse-6 v1.1 (Illumina) scl28809.4_20-S - -
          mouse-6 v1.1 (Illumina) scl0001058.1_1-S - -
          mouse-6 v1.1 (Illumina) scl0001236.1_24-S - -
          Illumina Sentrix mouse-6.v1 bead array 1050279 - -
          Illumina Sentrix mouse-6.v1 bead array 5220273 - -
          Illumina Sentrix mouse-6.v1 bead array 2680112 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl28809.4_20-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0001236.1_24-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0001058.1_1-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2680112 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1050279 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5220273 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1242767 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2704141 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1257069 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2704141 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1242767 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1257069 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline