| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0600006923.mm.1 |
TC0600000921.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600006923.mm.1 |
NM_001081266.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600006923.mm.1 |
NM_001081266.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600006932.mm.1 |
TC0600000921.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600006932.mm.1 |
NM_001081266.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600006932.mm.1 |
NM_001081266.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600006925.mm.1 |
TC0600000921.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600006925.mm.1 |
NM_001081266.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600006925.mm.1 |
NM_001081266.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600006924.mm.1 |
TC0600000921.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600006924.mm.1 |
NM_001081266.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600006924.mm.1 |
NM_001081266.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600006928.mm.1 |
TC0600000921.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600006928.mm.1 |
NM_001081266.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600006928.mm.1 |
NM_001081266.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600006926.mm.1 |
TC0600000921.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600006926.mm.1 |
NM_001081266.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600006926.mm.1 |
NM_001081266.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600006930.mm.1 |
TC0600000921.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600006930.mm.1 |
NM_001081266.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600006930.mm.1 |
NM_001081266.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600006927.mm.1 |
TC0600000921.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600006927.mm.1 |
NM_001081266.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600006927.mm.1 |
NM_001081266.1 |
Refseq |